Order Entry
Switzerland
ContactUsLinkComponent
Mixed calibration standard 5 for ICP
Mixed calibration standard 5 for ICP
Catalog # HONCE2098-100ML
CAS Number:  
Mixed calibration standard 5 for ICP
Catalog # HONCE2098-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Mixed Calibration Standard 5

Specifications

About this item

Certified Reference Material (CRM).

1000 mg/L Mg, 200 mg/L each of Sb, Tl, 100 mg/L B, 50 mg/L Ag in HNO₃ 5%