Combined Seven Anion Standard II for IC
Catalog # HONCE2030-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Combined Seven Anion Standard II
- Environmentally Preferable:
Specifications
About this item
Certified Reference Material (CRM).
200 mg/L PO43-, 100 mg/L each of Cl-, NO2-, Br-, NO3-, SO42-, 20 mg/L F- in H₂O