Combined Seven Anion Standard I for IC
Catalog # HONCE2029-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Combined Seven Anion Standard I
- Environmentally Preferable:
Specifications
About this item
Certified Reference Material (CRM).
150 mg/L of PO43-, SO42-; 100 mg/L of NO2-, Br-, NO3-; 30 mg/L Cl-; 20 mg/L F- in H₂O