Order Entry
Switzerland
ContactUsLinkComponent
Multi-element calibration standard 1 for ICP
Multi-element calibration standard 1 for ICP
Catalog # HONCE2026-100ML
CAS Number:  
Multi-element calibration standard 1 for ICP
Catalog # HONCE2026-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-Element Calibration Standard 1

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb in HNO₃ 5%