Order Entry
Switzerland
ContactUsLinkComponent
 
Multi-element calibration standard-4B for ICP
Catalog # HONCE2025-100ML
CAS Number:  
Multi-element calibration standard-4B for ICP
Catalog # HONCE2025-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃/tr. HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-Element Calibration Standard-4B

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr in HNO₃ 2%, tr HF