Specifications
- Pk:100 mg
- Source:Tail tendon
- Species:Rat
- Seq:05
- Environmentally Preferable:Low Manufacturing Impact
- Storage conditions:Stable for at least three months at 2 to 8 °C. Do not freeze.
- Environmental certification:ISO 14001 - Bedford
- Protein/peptide name:Collagen I
- Formulation:As a liquid in 0,02 N acetic acid. Typical protein concentration 3 - 4 mg/ml in standard preparation.
Specifications
About this item
Collagen I is found in most tissues and organs, but is most plentiful in dermis, tendon, and bone. Used as a thin coating in cell and tissue culture, it is often used to enhance cell attachment and proliferation. When applied as a gel it can be used to promote expression of cell-specific morphology and function. Collagen I is commonly used to culture endothelial cells, hepatocytes, muscle cells, and a variety of other cell types. The HC formulation of rat tail collagen I is used for three dimensional applications requiring a sturdy gel that provides a maximal 3D support matrix (guidelines are included with product).
Quality control: All preparations are quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma. Source material for human collagen I tested for hepatitis B antigen and HIV-1 antibody.