X-ray and metal detectable scoops, SteriWare®
Supplier: Sampling Systems
Certificates
About this item
The X-ray and metal detectable scoops are ideal for use in food and pharmaceutical production. They are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the powder in line with HACCP guidelines.
- Robust PP construction, provides excellent chemical resistance to many products
- Available in volumes from 30 to 2500 ml
- Full batch traceability
To ensure the cleanliness of the scoops they are manufactured and packed in a Cleanroom. They can be used as a single use disposable product straight from the bag or reused. The robust nature of these scoops means that they give many years of service.
Certifications: These PP scoops conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Packaging: Supplied individually bagged.
Frequently Bought Together