ContactUsLinkComponent
Avantor® Hichrom HI-DAI 2, GC Columns
Avantor® Hichrom HI-DAI 2, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-DAI 2, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI43-25-250-30
  • HI43-32-300-30
  • HI43-25-250-50
  • HI43-32-300-50
  • HI43-32-500-50
  • HI43-32-500-30
  • Phase name:
  • HI-DAI 2
  • HI-DAI 2
  • HI-DAI 2
  • HI-DAI 2
  • HI-DAI 2
  • HI-DAI 2
  • Phase composition:
  • Proprietary Phase developed for direct aqueous injections
  • Proprietary Phase developed for direct aqueous injections
  • Proprietary Phase developed for direct aqueous injections
  • Proprietary Phase developed for direct aqueous injections
  • Proprietary Phase developed for direct aqueous injections
  • Proprietary Phase developed for direct aqueous injections
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 320 °C
  • 320 °C
  • 320 °C
  • 320 °C
  • 320 °C
  • 320 °C