ContactUsLinkComponent
Avantor® Hichrom HI-BIODIESEL 110, GC Columns
Avantor® Hichrom HI-BIODIESEL 110, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-BIODIESEL 110, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI41-32-180-30
  • HI41-32-100-30
  • Phase name:
  • HI-BIODIESEL 110
  • HI-BIODIESEL 110
  • Phase composition:
  • Proprietary Phase developed for Biodiesel EN14110 analysis
  • Proprietary Phase developed for Biodiesel EN14110 analysis
  • Crossbond:
  • Yes
  • Yes
  • Temperature limit:
  • 380 °C
  • 380 °C