ContactUsLinkComponent
Avantor® Hichrom HI-5 Basic, GC Columns
 :  Avantor
undefined
Avantor® Hichrom HI-5 Basic, GC Columns
 :  Avantor
  • Catalog No:
  • HI03-32-150-15
  • HI03-32-100-60
  • HI03-32-150-50
  • HI03-32-150-30
  • HI03-32-150-60
  • HI03-25-100-50
  • HI03-32-100-15
  • HI03-25-100-60
  • HI03-32-100-50
  • HI03-32-100-30
  • HI03-25-025-15
  • HI03-25-025-50
  • HI03-25-025-30
  • HI03-25-050-15
  • HI03-25-025-60
  • HI03-25-050-50
  • HI03-25-050-30
  • HI03-25-100-15
  • HI03-25-050-60
  • HI03-53-100-30
  • HI03-25-100-30
  • HI03-53-100-15
  • HI03-53-100-60
  • HI03-53-100-50
  • Phase name:
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • HI-5 Basic
  • Phase composition:
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
  • Polarity:
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
Frequently Bought Together