ContactUsLinkComponent
Avantor® Hichrom HI-8 HT, GC Columns
Avantor® Hichrom HI-8 HT, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-8 HT, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI88-53-010-30
  • HI88-53-015-15
  • HI88-53-010-10
  • HI88-53-010-15
  • HI88-53-025-15
  • HI88-53-025-30
  • HI88-53-015-30
  • HI88-53-017-30
  • HI88-25-010-30
  • HI88-25-015-15
  • HI88-25-010-15
  • HI88-10-010-10
  • HI88-10-010-5
  • HI88-25-025-15
  • HI88-25-025-30
  • HI88-25-015-30
  • HI88-32-010-30
  • HI88-32-015-15
  • HI88-32-010-10
  • HI88-32-010-10G2
  • HI88-32-010-15
  • HI88-32-025-30
  • HI88-32-015-30
  • HI88-32-025-15
  • Phase name:
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • HI-8 HT
  • Phase composition:
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Proprietary Phase - high temperature
  • Polarity:
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Low-to-mid polarity stationary phase
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C