Avantor® Hichrom HI-DEX DMT Beta, GC Columns
Supplier: Avantor
Certificates
About this item
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI71-25-025-15
- HI71-25-025-25
- HI71-25-015-60
- HI71-25-025-60
- HI71-25-025-30
- HI71-25-025-50
- HI71-25-015-30
- HI71-25-015-50
- HI71-25-015-15
- HI83-10-010-10
- HI71-25-015-25
- HI83-10-010-5
- Phase name:
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- HI-DEX DMT Beta
- Phase composition:
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Dimethyl TBS Beta Cyclodexstrin based
- Polarity:
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Crossbond:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Temperature limit:
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C