ContactUsLinkComponent
Avantor® Hichrom HI-DEX DET Beta, GC Columns
Avantor® Hichrom HI-DEX DET Beta, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-DEX DET Beta, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI68-25-025-25
  • HI68-25-025-30
  • HI68-25-015-60
  • HI68-25-025-15
  • HI68-25-015-15
  • HI68-25-025-50
  • HI68-25-025-60
  • HI80-10-010-5
  • HI68-25-015-50
  • HI68-25-015-25
  • HI80-10-010-10
  • HI68-25-015-30
  • Phase name:
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • HI-DEX DET Beta
  • Phase composition:
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Diethyl TBS Beta Cyclodexstrin based
  • Polarity:
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Crossbond:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Temperature limit:
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C