Avantor® Hichrom HI-DEX DAC Beta, GC Columns
Anbieter: Avantor
Zertifikate
Über diesen Artikel
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.
Technische Daten
- Catalog No:
- HI66-25-015-50
- HI66-25-015-60
- HI66-25-025-15
- HI66-25-025-50
- HI78-10-010-5
- HI66-25-025-60
- HI66-25-025-25
- HI66-25-025-30
- HI78-10-010-10
- HI66-25-015-25
- HI66-25-015-30
- HI66-25-015-15
- Phase name:
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- HI-DEX DAC Beta
- Phase composition:
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Diacethyl TBS Beta Cyclodexstrin based
- Polarity:
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Chiral stationary phase
- Crossbond:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Temperature limit:
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
- 230 °C
Technische Daten
Frequently Bought Together