ContactUsLinkComponent
Avantor® Hichrom HI-SOLVE 1, GC Columns
 :  Avantor
undefined
Avantor® Hichrom HI-SOLVE 1, GC Columns
 :  Avantor
  • Catalog No:
  • HI62-25-025-30
  • HI62-32-025-50
  • Phase name:
  • HI-SOLVE 1
  • HI-SOLVE 1
  • Phase composition:
  • Proprietary Phase developed for complex solvent mix
  • Proprietary Phase developed for complex solvent mix
  • Crossbond:
  • Stabilised
  • Stabilised
Frequently Bought Together