ContactUsLinkComponent
Avantor® Hichrom HI-1 PONA, GC Columns
Avantor® Hichrom HI-1 PONA, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-1 PONA, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI57-25-100-150
  • HI57-20-050-100
  • HI57-25-050-100
  • HI57-20-050-50
  • Phase name:
  • HI-1 PONA
  • HI-1 PONA
  • HI-1 PONA
  • HI-1 PONA
  • Phase composition:
  • 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - optimized for hydrocarbon analysis
  • 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - optimized for hydrocarbon analysis
  • 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - optimized for hydrocarbon analysis
  • 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - optimized for hydrocarbon analysis
  • Polarity:
  • Apolar stationary phase
  • Apolar stationary phase
  • Apolar stationary phase
  • Apolar stationary phase
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C