ContactUsLinkComponent
Avantor® Hichrom HI-BIODIESEL 105, GC Columns
Avantor® Hichrom HI-BIODIESEL 105, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-BIODIESEL 105, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI40-32-010-10
  • HI40-53-050-5
  • HI40-32-010-15
  • Phase name:
  • HI-BIODIESEL 105
  • HI-BIODIESEL 105
  • HI-BIODIESEL 105
  • Phase composition:
  • Proprietary Phase developed for Biodiesel EN14105 analysis
  • Proprietary Phase developed for Biodiesel EN14105 analysis
  • Proprietary Phase developed for Biodiesel EN14105, D6584 analysis
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 380 °C
  • 380 °C
  • 380 °C