Avantor® Hichrom HI-10, GC Columns
Supplier: Avantor
Certificates
About this item
Application specific highly polar phase, ideal for detailed cis/trans FAMEs separation.
- Phase composition: 100% Cyanopropyl Polysiloxane
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HIGCHI262502050/5
- HI26-25-025-120
- HIGCHI26-20-020-30
- HI26-32-020-30
- HI26-32-020-105
- HI26-32-020-60
- HI26-32-020-50
- HI26-32-025-105
- HI26-32-025-100
- HI26-32-025-50
- HI26-20-020-105
- HI26-32-025-30
- HI26-20-020-100
- HI26-18-010-20
- HI26-20-020-50
- HI26-32-025-60
- HI26-20-010-10
- HI26-20-020-40
- HI26-20-020-60
- HI26-18-010-10
- HI26-18-010-40
- HI26-20-010-20
- HI26-20-020-20
- HI26-32-020-100
- HI26-25-020-105
- HI26-25-020-100
- HI26-20-010-105
- HI26-25-020-50
- HI26-25-020-30
- HI26-20-010-100
- HI26-20-010-50
- HI26-25-025-100
- HI26-25-020-60
- HI26-20-010-40
- HI26-25-025-30
- HI26-20-010-60
- HI26-25-025-105
- HI26-25-025-60
- HI26-25-025-50
- Phase name:
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- Phase composition:
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- Crossbond:
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Temperature limit:
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- USP code:
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48