Orders LinkContactUsLinkComponent
Mobile container spill kit
Mobile container spill kit
Supplier:   Brady
Mobile container spill kit
Supplier:  Brady
Certificates
Mobile container spill kit
Mobile container spill kit
2 Options Available Below
Product Details & Documents
These wheeled spill kits are comes in a polyethylene container with hinged lid. Designed for medium sized spills of industrial liquids including oils, water, solvents, coolants and chemical hazardous liquids.
  • Yellow in colour
Container is weather-resistant and protects the sorbents against dirt, moisture and damage. Hinged lid provides easy access to the products. Wheels allow excellent mobility indoors and outdoors.
Product Family Options
Product Information
  • Absorbency
    W×D×H
    Availability
    Price
  • BRDY813882
    Mobile container maintanence kit
    Mobile container maintanence kit
    124 L
    500×540×900 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 644.00 SFr.
    644.00 SFr.
    1 * 1 KIT
       644.00 SFr.
  • BRDY813883
    Mobile container chemical kit
    Mobile container chemical kit
    128 L
    500×540×900 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 688.00 SFr.
    688.00 SFr.
    1 * 1 KIT
       688.00 SFr.