ContactUsLinkComponent
Labels for thermal transfer printers, type B-438
Labels for thermal transfer printers, type B-438
Catalog # BRDY030182
Supplier:  Brady
undefined
Labels for thermal transfer printers, type B-438
Catalog # BRDY030182
Supplier:  Brady
Specifications
  • Type:
    B-438
  • Colour:
    Silver
  • Description:
    Fraud resistant labels, checkerboard pattern
  • Dimensions:
    50×25 mm
  • Material:
    Polyester
  • Model:
    THT-17-438-3
  • Packed:
    3000 labels/roll
  • Pk:
    1 Roll
  • Adhesive:
    Acrylic
Specifications
About this item
  • Fuels or oils resistant
  • High and low temperatures resistant
  • Solvents or chemicals resistant
  • Fraud resistant
Certifications: UL recognised.