Labels for thermal transfer printers, type B-473
Supplier: Brady
Certificates
About this item
Glossy, electrostatically dissipative polyester labels for printed circuit boards.
- Fuels or oils resistant
- High and low temperatures resistant
- Solvents or chemicals resistant
Certifications: UL recognised, CSA accepted, meets surface resistances within the recommended range for absorbent ESD packaging materials as defined by ANSI/ESD S541-2008.