ContactUsLinkComponent
Oil drum top covers
Oil drum top covers
Catalog # BRDY813784
Supplier:  Brady
Oil drum top covers
Catalog # BRDY813784
Supplier:  Brady
Specifications
  • Description:
    Drum top covers, Ø: 560 mm, white
  • Pk:
    25
Specifications
About this item
Oil sorbent drum top cover with pre-cut holes that absorbs oil and petroleum based fluids.
This helps to keep drum top, work floor and surrounding storage area neat and clean by preventing drips from reaching the floor.