Order Entry
Switzerland
ContactUsLinkComponent
 
Thermal transfer ribbon for printer, R7964 series
  BRDY237588
 :  Brady
undefined
Thermal transfer ribbon for printer, R7964 series
  BRDY237588
 :  Brady

 

  • Colour:
    Black
  • Description:
    Thermal transfer printer ribbon, R7964 series, resin formulation, width 110 mm
  • Length:
    100 m
  • Environmentally Preferable:
  • Pk:
    1
  • For:
    BP1244, BP1344

 

 

If the R-7964 print ribbon is printed on the correct Brady label, this guarantees good solvent resistance and razor sharp print quality.

 : One print ribbon/box.