Chemical resistant overalls, AlphaTec® 2000 Standard, Model 111
Supplier: Ansell
Certificates
About this item
Superior breathable microporous laminate technology, offering Type 5/6 protection.
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Comfortable moisture vapor permeable fabric helps reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5
- Low linting to help reduce the risk of cross contamination in critical areas
- Silicone-free, critical in spray painting applications
Coveralls feature a 3-piece hood, 2-way front zipper with resealable storm flap, and finger loops. Elasticated hood, waist, wrists, and ankles. Garments are manufactured from non-woven microporous polyethylene laminate, with bound seams.
Recommended for cleanroom cleaning and preparation, production line support and maintenance, maintenance of plant and machines, and veterinary services.
Certifications: Type 5-B; Type 6-B; EN 1073-2; EN 14126; DIN 32781; EN 1149-5; Category III
Packaging: Individually packaged; 40 pieces/carton.
Specifications
- Catalog No:
- ANSEGR20T-00111-06
- ANSEGR20T-00111-05
- ANSEGR20T-00111-03
- ANSEGR20T-00111-07
- ANSEGR20T-00111-02
- ANSEGR20T-00111-04
- ANSEWH20T-00111-08
- ANSEWH20T-00111-09
- Model:
- 111
- 111
- 111
- 111
- 111
- 111
- 111
- 111
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection