Über diesen Artikel
Lightweight, breathable microporous laminate technology, offering Type 5/6 protection.
- Protects against low hazard liquid spray and fine particulates
- Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5 and AATCC 76
- Low linting to help reduce the risk of cross contamination in critical areas
- Silicone free, critical in spray painting applications
Designed for workers in warm environments to provide an optimum balance between comfort and protection. Garments are manufactured from microporous polyethylene laminate nonwoven with polypropylene nonwoven with anti-static treatment (back, underarm, and hood). These overalls feature a 3-piece hood, navy blue binding with SMS hood, full back and leg panels. Elasticated hood, waist, ankles and wrists, 2-way front zipper with resealable storm flap, finger loops. Bound seams.
Recommended for cleanroom cleaning and preparing, body painting including surface inspection, production line support and maintenance, cleaning and maintenance of plant and machines, fibre glass resin applications, powder handling, and general purpose auto aftermarket applications.
Technische Daten
- Catalog No:
- ANSEWN18B-00195-03
- ANSEWN18B-00195-02
- ANSEWN18B-00195-07
- ANSEWN18B-00195-04
- ANSEWN18B-00195-06
- ANSEWN18B-00195-05
- ANSEWN18B-00195-08
- ANSEWN18B-00195-09
- Model:
- 195
- 195
- 195
- 195
- 195
- 195
- 195
- 195
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection