Tetraethylammonium hexafluorophosphate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Tetraethylammoniumhexafluorophosphate
CAS Number:429-07-2
:
Formula:C₈H₂₀F₆NP
MDL Number:MFCD00043179
:275.22 g/mol
Storage Temperature:Ambient
EINECS:207-056-7
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together