Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bismuth(III) nitrate basic

Bismuth(III) nitrate basic

Glass bottle for solids, 50 g
Catalog # ROTH4447.1
Oxidizing
Irritant
Marketsource
Supplier:   Roth Carl
Bismuth(III) nitrate basic
Catalog # ROTH4447.1
Supplier:  Roth Carl
Marketsource
CAS Number:  1304-85-4
Bismuth(III) nitrate basic

Glass bottle for solids, 50 g

CAS Number:  1304-85-4
Price
SFr. 68.30
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
  • CAS Number:1304-85-4
  • Formula:Bi₅O(OH)₉(NO₃)₄
  • MDL Number:MFCD00064844
  • Molecular Weight:1461.99 g/mol
  • Density:4.928 g/cm³ (20 °C)
  • EINECS:215-136-8
  • UN:1477
  • ADR:5.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications