Quartz
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Silicium dioxide, Silicium (IV) oxide, Silicon dioxide, Sand, white quartz
- CAS Number:14808-60-7
- Formula:SiO₂
- MDL Number:MFCD00217788
- Molecular Weight:60.08 g/mol
- Boiling Pt:2230 °C (1013 hPa)
- Melting Pt:1427 °C
- Density:2.622…2.645 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:238-878-4
2 Options Available Below
Product Details & Documents
Synonyms: Silicium dioxide, Silicium (IV) oxide, Silicon dioxide, Sand, white quartz
- CAS Number:14808-60-7
- Formula:SiO₂
- MDL Number:MFCD00217788
- Molecular Weight:60.08 g/mol
- Boiling Pt:2230 °C (1013 hPa)
- Melting Pt:1427 °C
- Density:2.622…2.645 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:238-878-4



