Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Quartz

Marketsource
Supplier:   Roth Carl
Quartz
Supplier:  Roth Carl
Marketsource
Certificates
Quartz

2 Options Available Below

Product Details & Documents
Synonyms:  Silicium dioxide, Silicium (IV) oxide, Silicon dioxide, Sand, white quartz
  • CAS Number:14808-60-7
  • Formula:SiO₂
  • MDL Number:MFCD00217788
  • Molecular Weight:60.08 g/mol
  • Boiling Pt:2230 °C (1013 hPa)
  • Melting Pt:1427 °C
  • Density:2.622…2.645 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:238-878-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - SFr. 27.50
    SFr. 27.50
    1 * 2,5 kg
       SFr. 27.50

    Specifications:
    Marketsource
    Marketsource
  • 20 kg
    Log in for availability
    Log in for availability
    1 * 20 kg
    1 * 20 kg - SFr. 97.10
    SFr. 97.10
    1 * 20 kg
       SFr. 97.10

    Specifications:
    Marketsource
    Marketsource