Order Entry
Switzerland
ContactUsLinkComponent
Aluminium sulphate octadecahydrate, TECHNICAL
Corrosive
Aluminium sulphate octadecahydrate, TECHNICAL
Catalog # 21068.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Aluminium sulphate octadecahydrate, TECHNICAL
Catalog # 21068.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :666.43 g/mol
Melting Pt:86 °C
Density:1.69 g/cm³ (20 °C)
EINECS:233-135-0
 :GHS05
CAS Number:7784-31-8
Formula:Al₂(SO₄)₃·18H₂O
MDL Number:MFCD00149136