

Kaliumsulfid (K2Sx), EMSURE® zur Analyse, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:37199-66-9
:39365-88-3
Formula:K₂(S)n
MDL Number:MFCD00148074
:110.26 g/mol
Melting Pt:471 °C
Density:1,65 g/cm³ (20 °C)
EINECS:253-390-1
UN:3262
ADR:8,II
:M-Clarity™ Qualitätsstufe = MQ300
:GHS05
:GHS09
Storage Temperature:Raumtemperatur
Specification Test Results
- Solubility:500 g/l (20 °C)
- Melting Point:200 - 250 °C
- Bulk Density:1000 - 1200 kg/m3
- Density:1.65 g/cm3 (20 °C)
- pH:13 (10 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot Numberexternalurl.pdf ProductPage_105134
Product specification
msds_105134
SDS
Frequently Bought Together