Sodium hexametaphosphate, soluble, GPR RECTAPUR®
Catalog # 301475H
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Assay (as P₂O₅):68-72 %
- Substances insoluble in water:Max. 0.1 %
- pH range:6.2 - 7.4
- Hg (Mercury):Max. 5 ppm
- Pb (Lead):Max. 5 ppm
Specification Test Results
About this item
:
:611.77 g/mol
Melting Pt:610 °C
Density:1.25 g/cm³ (20 °C)
EINECS:233-343-1
CAS Number:10124-56-8
Formula:Na₆P₆O₁₈
MDL Number:MFCD00136045