Order Entry
Switzerland
ContactUsLinkComponent
Potassium chloride 99.0-100.5%, granular, AR® ACS, Macron Fine Chemicals™
Potassium chloride 99.0-100.5%, granular, AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Potassium chloride 99.0-100.5%, granular, AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay (as KCl) (dried basis):
    99.0 - 100.5 %
  • ACS - Barium (Ba):
    Passes Test
  • ACS - Bromide (Br):
    ≤0.01 %
  • ACS - Calcium (Ca):
    ≤0.002 %
  • ACS - Chlorate and Nitrate (as NO₃):
    ≤0.003 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iodide (I):
    ≤0.002 %
  • ACS - Iron (Fe):
    ≤3 ppm
  • ACS - Magnesium (Mg):
    ≤0.001 %
  • ACS - pH of 5% Solution at 25°C:
    5.4 - 8.6
  • ACS - Phosphate (PO₄):
    ≤5 ppm
  • ACS - Sodium (Na):
    ≤0.005 %
  • ACS - Sulfate (SO₄):
    ≤0.001 %
  • ACS - Loss on Drying at 105°C:
    ≤1 %

Specification Test Results

Related Information

Frequently Bought Together