


Chloramin T (Natriumsalz) Trihydrat ACS, Reag. Ph. Eur. analytisches Reagens, Supelco®
Catalog # 1.02426.0250
Supplier: Merck
CAS Number:
Specifications
- Packung:Kunststoffflasche
- VE:250 g
- Seq:3545
Specifications
Specification Test Results
- Solubility:150 g/l (25 °C)
- Molar Mass:281.69 g/mol
- Bulk Density:540 - 680 kg/m3
- Flash Point:192 °C
- pH:8 - 10 (50 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
About this item
:
Merck Index:13,02085
Melting Pt:167…170 °C
Density:1.43 g/cm³ (20 °C)
EINECS:204-854-7
ADR:6.1 (8),II
:M-Clarity™ quality level = MQ100
Flash Pt:192 °C (closed cup)
:281.69 g/mol
UN:2928
:GHS05
:GHS08
:GHS07
Storage Temperature:Kühlschrank
CAS Number:7080-50-4
Formula:CH₃C₆H₄SO₂NClNa·3H₂O
MDL Number:MFCD00149066