Wipe rolls, WypAll® L10 compact food and hygiene
KIMB7225
- Packed:165 sheets/roll
- Colour:Blue
- Description:Compact roll, 1-ply
- L×W:460×240 mm
- Seq:01
- Pk:24 Roll
Compact disposable paper wipes are 1-ply and designed for light-duty wiping in areas such as food preparation and service surfaces.
- Compact roll design is small, portable, and easy to handle
- Ease of use helps to support hygiene standards and compliance while also saving space
- WypAll® wiping paper is designed to be used once with detergent or disinfectant
These dry cleaning wipes have been tested as compatible for use in spray and wipe application with peracetic acid, hydrogen peroxide, and quats (quaternary ammonium compounds). Spray and wiping compatibility helps to ensure workers can wipe own work spaces, vehicles, desks, seating, and food prep areas before and after use. Spray and wipe also helps to enable production workers to clean down/sanitise individual workstations before hand-off to next shift. For environments where fully saturated wiping is a requirement, these WypAll® rolls are tested as compatible with soap/detergent.