Order Entry
Switzerland
ContactUsLinkComponent
Phenol 99.0-100.5% (by anhydrous basis), crystals USP, Macron Fine Chemicals™
Toxic
Corrosive
Health
Phenol 99.0-100.5% (by anhydrous basis), crystals USP, Macron Fine Chemicals™
Certificates
undefined
Toxic
Corrosive
Health
Phenol 99.0-100.5% (by anhydrous basis), crystals USP, Macron Fine Chemicals™
Certificates

Specification Test Results

  • GMP Manufactured Product:
    99.0 - 100.5%
  • Meets U.S.P Requirements:
    ≤2.0 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (C₆H₅OH) (anhydrous basis):
    99.0 - 100.5 %
  • USP - Appearance of Melted Sample:
    Passes Test
  • USP - Benzene:
    ≤2.0 ppm
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Clarity of Solution and Reaction:
    Passes Test
  • USP - Congealing Temperature:
    ≥39 °C
  • USP - Nonvolatile Residue:
    ≤0.05 %
  • USP - Water (H₂O)(by Karl Fischer titrn):
    ≤0.5 %

Specification Test Results

Related Information

Frequently Bought Together