4-Nitrophenol
Glass bottle for solids, 100 mg
Catalog # PROOMM0042.03
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: 1-Hydroxy-4-nitrobenzene, p-Nitrophenol
- CAS Number:100-02-7
- Formula:C₆H₅NO₃
- MDL Number:MFCD00007331
- Molecular Weight:139.11 g/mol
- Merck Index:13,06654
- Boiling Pt:279 °C (1013 hPa)
- Melting Pt:110…114 °C
- Density:1.48 g/cm³ (20 °C)
- Flash Pt:169 °C
- Storage Temperature:Ambient
- EINECS:202-811-7
- UN:1663
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



