2-Nitro-p-anisidine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-96-8
:
Formula:C₇H₈N₂O₃
MDL Number:MFCD00007152
:168.15 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:123…126 °C
Flash Pt:≥ 200 °C
Storage Temperature:Ambient
EINECS:202-547-2
UN:2811
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:Orange Crystalline powder
- Infrared spectrum:Conforms
- Melting point:122 to 126 °C
- HPLC:≥96.0 %
- Elemental Analysis:18.13 to 19.07 % N
- Iron (Fe):≤5 ppm
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together