Order Entry
Switzerland
Orders LinkContactUsLinkComponent
4-Nitro-o-anisidine 99%

4-Nitro-o-anisidine 99%

Irritant
4-Nitro-o-anisidine 99%
Certificates
4-Nitro-o-anisidine 99%

3 Options Available Below

Product Details & Documents
Synonyms:  2-Methoxy-4-nitroaniline, 2-Amino-5-nitroanisole
  • CAS Number:97-52-9
  • Formula:C₇H₈N₂O₃
  • MDL Number:MFCD00007363
  • Molecular Weight:168.15 g/mol
  • Boiling Pt:364 °C (1013 hPa)
  • Melting Pt:139…142 °C
  • Density:1,2112 g/cm³ (156 °C)
  • Storage Temperature:Ambient
  • EINECS:202-588-6

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow to orange Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    139 to 142 °C
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 33.00
    SFr. 33.00
    1 * 5 g
       SFr. 33.00

  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 46.40
    SFr. 46.40
    1 * 100 g
       SFr. 46.40

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 137.00
    SFr. 137.00
    1 * 500 g
       SFr. 137.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.