Order Entry
Switzerland
ContactUsLinkComponent
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) ≥98.0%
Irritant
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) ≥98.0%
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB) ≥98.0%
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98.0 %
  • Appearance:
    Yellow crystalline powder

Specification Test Results

Related Information

Frequently Bought Together