ContactUsLinkComponent
Industrial gloves, HyFlex® 11-931
Industrial gloves, HyFlex® 11-931
Supplier:  Ansell
Certificates
undefined
Industrial gloves, HyFlex® 11-931
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE11-931/10
  • ANSE11-931/8
  • ANSE11-931/6
  • ANSE11-931/7
  • ANSE11-931/11
  • ANSE11-931/9
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Liner material:
  • Dyneema®
  • Dyneema®
  • Dyneema®
  • Dyneema®
  • Dyneema®
  • Dyneema®
  • Finish:
  • Foam nitrile palm coating
  • Foam nitrile palm coating
  • Foam nitrile palm coating
  • Foam nitrile palm coating
  • Foam nitrile palm coating
  • Foam nitrile palm coating
  • Gauge:
  • 18
  • 18
  • 18
  • 18
  • 18
  • 18
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +