Order Entry
Switzerland
ContactUsLinkComponent
Valethamate bromide ≥98.0% (by HPLC, titration analysis)
 
Valethamate bromide ≥98.0% (by HPLC, titration analysis)
  TCIAV0063-1G
 :  TCI
 :  
undefined
 
Valethamate bromide ≥98.0% (by HPLC, titration analysis)
  TCIAV0063-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

 

 

 :
 :386.37 g/mol
 :14,09906
 :201-977-8
 :GHS07
 :90-22-2
 :C₁₉H₃₂BrNO₂
 :MFCD00031617