Tissue-clearing reagent, CUBIC-B for animals
Catalog # TCIAT3780-100ML
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mL
- Description:Tissue-Clearing reagent, CUBIC-B for Animals
Specifications
About this item
Specifications
Appearance: Colorless to Almost colorless clear liquid
Functionality test: to pass test (Remaining fluorescence strength)
Decalcification test: max. 0.3 (590 to 600 nm)