Order Entry
Switzerland
ContactUsLinkComponent
Tissue-clearing reagent, CUBIC-L for animals
Tissue-clearing reagent, CUBIC-L for animals
  TCIAT3740-100ML
 :  TCI
 :  
Tissue-clearing reagent, CUBIC-L for animals
  TCIAT3740-100ML
 :  TCI
 :  

 

  • Pk:
    100 mL
  • Description:
    Tissue-Clearing reagent, CUBIC-L for Animals

 

 

Specifications
Appearance: Colorless to Almost colorless clear liquid
Functionality test: to pass test (Remaining fluorescence strength)