Order Entry
Switzerland
ContactUsLinkComponent
Tripropylene glycol monomethyl ether (mixture of isomer) ≥93.0% (by GC)
Tripropylene glycol monomethyl ether (mixture of isomer) ≥93.0% (by GC)
Supplier:  TCI
Certificates
Tripropylene glycol monomethyl ether (mixture of isomer) ≥93.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 93.0 % (total of isomers)

Specification Test Results

Frequently Bought Together