
MOPS-Na ≥99%, high purity
Catalog # E413-250G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Abs.@260 nm (0.1 M, Water):<0.03
- DNase:none detected
- Heavy Metals:<0.005%
- Appearance:White powder
- Loss on Drying:< 1%
- pH (0.1 M, Water) @25 °C:10 – 11
- pKa (20 °C):7 – 7.4
- Protease:none detected
- Purity:> 99%
- RNase:none
- Solubility (0.1 M, Water):Pass
Specification Test Results
About this item
:
:231.25 g/mol
Melting Pt:277 °C
EINECS:428-420-3
:GHS07
Storage Temperature:Ambient
CAS Number:71119-22-7
Formula:C₇H₁₄NNaO₄S
MDL Number:MFCD00064350