Phytantriol (mixture of isomers) ≥95.0% (by GC)
TCIAP1674-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 % (total of isomers)
- Purity (Hydroxyl Value):min. 98.0 %
:
:330.55 g/mol
:14,07386
:199°C
:277-923-2
:74563-64-7
:C₂₀H₄₂O₃