
Octadecenylsuccinic anhydride (mixture of isomers) ≥93.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:28777-98-2
:
Formula:C₂₂H₃₈O₃
MDL Number:MFCD00014545
:350.54 g/mol
EINECS:249-210-6
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 93.0 %
- Purity (Morpholine Method):min. 90.0 %
- Melting point:67.0 to 73.0 deg-C
Specification Test Results
Frequently Bought Together