Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, 3-Bromobenzaldehyde
Supelco® Organic Reference Standard, 3-Bromobenzaldehyde
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, 3-Bromobenzaldehyde
Supplier:  Merck

Specifications

  • CAS No.:
    3132-99-8
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00003345
  • M-Clarity quality level:
    MQ100
  • Formula:
    BrC₆H₄CHO
  • Molecular weight:
    185,02
  • Flash point:
    110 °C - Closed cup

Specifications

Frequently Bought Together