Order Entry
Switzerland
ContactUsLinkComponent
 
Sodium-2-naphthol-7-sulphonate hydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0036-25G
Supplier:  TCI
CAS Number:  
undefined
Sodium-2-naphthol-7-sulphonate hydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0036-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Red powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (Ion exchange titration):
    min. 98.0 % (calcd.on anh.substance) (after deduct NaCl content)
  • Water:
    max. 8.0 %

Specification Test Results

About this item

 :
 :246.21 g/mol
EINECS:205-200-3
CAS Number:135-55-7
Formula:C₁₀H₈O₄S
MDL Number:MFCD00021625